Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGGTAGATCCCTGGTTTCTGG[A/G]AGGAGGAGGATAACAGGGACAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A24 - solute carrier family 22 member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136506.2 | 1987 | Silent Mutation | CTC,CTT | L,L 515 | NP_001129978.2 | |
NM_173586.2 | 1987 | Intron | NP_775857.2 | |||
XM_011544967.2 | 1987 | Intron | XP_011543269.1 |