Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACCCTGGGGTATCACCCCATT[A/G]AGCAGGATGCAGATGCCTGCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607582 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC22A6 - solute carrier family 22 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004790.4 | 1566 | Silent Mutation | CTC,CTT | L,L 411 | NP_004781.2 | |
NM_153276.2 | 1566 | Silent Mutation | CTC,CTT | L,L 411 | NP_695008.1 | |
NM_153277.2 | 1566 | Silent Mutation | CTC,CTT | L,L 411 | NP_695009.1 | |
NM_153278.2 | 1566 | Silent Mutation | CTC,CTT | L,L 411 | NP_695010.1 | |
XM_017018562.1 | 1566 | Silent Mutation | CTC,CTT | L,L 412 | XP_016874051.1 |
Set Membership: |
DME Validated Inventoried |