Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACTCCAAATTCAAGGCTTTTC[C/T]CATTTTCTGCTTGTGAAACATGTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610966 MIM: 610937 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTO PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FTO - fat mass and obesity associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080432.2 | Intron | NP_001073901.1 | ||||
XM_011523313.2 | Intron | XP_011521615.1 | ||||
XM_011523314.2 | Intron | XP_011521616.1 | ||||
XM_011523315.2 | Intron | XP_011521617.1 | ||||
XM_011523316.2 | Intron | XP_011521618.1 | ||||
XM_017023654.1 | Intron | XP_016879143.1 | ||||
XM_017023655.1 | Intron | XP_016879144.1 | ||||
XM_017023656.1 | Intron | XP_016879145.1 | ||||
XM_017023657.1 | Intron | XP_016879146.1 | ||||
XM_017023658.1 | Intron | XP_016879147.1 |
RPGRIP1L - RPGRIP1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |