Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAAAGAATAGACATAAAATTCA[A/T]CTGTTCATTGTAGAGTGCTTCCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613344 | ||||||||||||||||||||
Literature Links: |
KIAA1549 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1549 - KIAA1549 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164665.1 | 8958 | UTR 3 | NP_001158137.1 | |||
NM_020910.2 | 8958 | UTR 3 | NP_065961.2 | |||
XM_011516442.2 | 8958 | UTR 3 | XP_011514744.1 |