Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGCCTTTAACATGTACCCCCG[A/G]AAGCTAAAATAAAAGTTAAGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609962 | ||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4E PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CLEC4E - C-type lectin domain family 4 member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014358.3 | 1626 | UTR 3 | NP_055173.1 | |||
XM_011520614.2 | 1626 | Intron | XP_011518916.1 | |||
XM_011520615.1 | 1626 | Intron | XP_011518917.1 |
Set Membership: |
HapMap |