Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGTAATTTTAAAGGATTCTCTG[G/T]TTTTTCCCTTCTATTATGTTTCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HN1 - hematological and neurological expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002032.2 | Intron | NP_001002032.1 | ||||
NM_001002033.2 | Intron | NP_001002033.1 | ||||
NM_001288609.1 | Intron | NP_001275538.1 | ||||
NM_001288610.1 | Intron | NP_001275539.1 | ||||
NM_001288611.1 | Intron | NP_001275540.1 | ||||
NM_016185.3 | Intron | NP_057269.1 |