Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGTCCTGTAGTTGTTTCTCTGA[A/G]GTTACATTTCGTCCATTTTCCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300440 MIM: 300683 | ||||||||||||||||||||
Literature Links: |
NKRF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKRF - NFKB repressing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEPT6 - septin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015129.5 | 3734 | Intron | NP_055944.2 | |||
NM_145799.3 | 3734 | Intron | NP_665798.1 | |||
NM_145800.3 | 3734 | UTR 3 | NP_665799.1 | |||
NM_145802.3 | 3734 | UTR 3 | NP_665801.1 | |||
XM_005262400.3 | 3734 | UTR 3 | XP_005262457.1 | |||
XM_006724748.2 | 3734 | UTR 3 | XP_006724811.1 | |||
XM_006724750.2 | 3734 | UTR 3 | XP_006724813.1 | |||
XM_011531317.2 | 3734 | UTR 3 | XP_011529619.1 | |||
XM_011531318.2 | 3734 | UTR 3 | XP_011529620.1 |