Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCGGGCGCAGCATGACTGCCG[C/T]CGGCGTGCAGGTAGCCGGCGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182098 | ||||||||||||||||||||
Literature Links: |
SCTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCTR - secretin receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM37 - transmembrane protein 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183240.2 | 45 | Missense Mutation | GCC,GTC | A,V 4 | NP_899063.2 | |
XM_005263598.4 | 45 | Intron | XP_005263655.1 | |||
XM_006712300.3 | 45 | Intron | XP_006712363.1 | |||
XM_011510659.2 | 45 | Intron | XP_011508961.1 |