Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601936 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAWR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PAWR - pro-apoptotic WT1 regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002583.2 | 1171 | Silent Mutation | GTG,GTT | V,V 335 | NP_002574.2 | |
XM_006719435.2 | 1171 | Silent Mutation | GTG,GTT | V,V 335 | XP_006719498.1 | |
XM_006719436.3 | 1171 | Intron | XP_006719499.1 | |||
XM_017019377.1 | 1171 | Silent Mutation | GTG,GTT | V,V 335 | XP_016874866.1 | |
XM_017019378.1 | 1171 | Intron | XP_016874867.1 | |||
XM_017019379.1 | 1171 | Intron | XP_016874868.1 |