Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAGGCAGGAAATCAGGCCAGA[A/G]AGGAAGGCTGGGGCTGGTCTGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615123 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD13A - ankyrin repeat domain 13A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C12orf76 - chromosome 12 open reading frame 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207435.2 | Intron | NP_997318.1 | ||||
XM_017019297.1 | Intron | XP_016874786.1 |