Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGTTACTGGGCCTGTCTCATT[C/G]TCACTTTTGCCACCTGCCAATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610648 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CUX2 - cut like homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015267.3 | Intron | NP_056082.2 | ||||
XM_011538061.2 | Intron | XP_011536363.1 | ||||
XM_011538063.2 | Intron | XP_011536365.1 | ||||
XM_011538069.2 | Intron | XP_011536371.1 | ||||
XM_011538070.2 | Intron | XP_011536372.1 | ||||
XM_017019080.1 | Intron | XP_016874569.1 | ||||
XM_017019081.1 | Intron | XP_016874570.1 |
Set Membership: |
HapMap |