Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGTCCTACAGGCCTAGTGGACT[A/C]TGAGCGGAGAATGTTATGACCACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602287 | ||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034025.1 | Intron | NP_001029197.1 | ||||
NM_006817.3 | Intron | NP_006808.1 | ||||
XM_017018720.1 | Intron | XP_016874209.1 |
TMEM116 - transmembrane protein 116 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193453.1 | Intron | NP_001180382.1 | ||||
NM_001193531.1 | Intron | NP_001180460.1 | ||||
NM_001294314.1 | Intron | NP_001281243.1 | ||||
NM_138341.2 | Intron | NP_612350.1 | ||||
XM_006719678.3 | Intron | XP_006719741.1 | ||||
XM_011538946.1 | Intron | XP_011537248.1 | ||||
XM_011538947.1 | Intron | XP_011537249.1 | ||||
XM_011538948.2 | Intron | XP_011537250.1 | ||||
XM_011538949.2 | Intron | XP_011537251.1 | ||||
XM_011538950.1 | Intron | XP_011537252.1 | ||||
XM_011538951.2 | Intron | XP_011537253.1 | ||||
XM_011538953.1 | Intron | XP_011537255.1 | ||||
XM_011538955.1 | Intron | XP_011537257.1 | ||||
XM_011538957.1 | Intron | XP_011537259.1 | ||||
XM_011538958.1 | Intron | XP_011537260.1 | ||||
XM_017020187.1 | Intron | XP_016875676.1 | ||||
XM_017020188.1 | Intron | XP_016875677.1 | ||||
XM_017020189.1 | Intron | XP_016875678.1 | ||||
XM_017020190.1 | Intron | XP_016875679.1 | ||||
XM_017020191.1 | Intron | XP_016875680.1 | ||||
XM_017020192.1 | Intron | XP_016875681.1 | ||||
XM_017020193.1 | Intron | XP_016875682.1 | ||||
XM_017020194.1 | Intron | XP_016875683.1 |