Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGATCCTCTCACCTTTCATCTT[C/T]TCCTGTGTCTACAGGTGGTCAGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603447 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HRK - harakiri, BCL2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003806.2 | 2943 | Intron | NP_003797.1 | |||
XM_017020146.1 | 2943 | UTR 3 | XP_016875635.1 |
RNFT2 - ring finger protein, transmembrane 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |