Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACTTCTGACATGCCTGTGATG[C/T]TTATTCTCAATTTTTCAGGTGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605571 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIWIL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIWIL1 - piwi like RNA-mediated gene silencing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190971.1 | Intron | NP_001177900.1 | ||||
NM_004764.4 | Intron | NP_004755.2 | ||||
XM_011539002.2 | Intron | XP_011537304.1 | ||||
XM_011539003.2 | Intron | XP_011537305.1 | ||||
XM_011539004.2 | Intron | XP_011537306.1 | ||||
XM_011539005.1 | Intron | XP_011537307.1 | ||||
XM_011539006.2 | Intron | XP_011537308.1 | ||||
XM_017020229.1 | Intron | XP_016875718.1 |
Set Membership: |
HapMap |