Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHH - sonic hedgehog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000193.3 | 3846 | Silent Mutation | CTC,CTG | L,L 299 | NP_000184.1 | |
NM_001310462.1 | 3846 | Intron | NP_001297391.1 | |||
XM_011516479.2 | 3846 | Silent Mutation | CTC,CTG | L,L 212 | XP_011514781.1 | |
XM_011516480.2 | 3846 | Silent Mutation | CTC,CTG | L,L 212 | XP_011514782.1 |