Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCAAGTTACATGATTTTAGCT[A/G]TGAAAGGCAATTTTGCTGTATTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602867 MIM: 180661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFBP7 - insulin like growth factor binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253835.1 | Intron | NP_001240764.1 | ||||
NM_001553.2 | Intron | NP_001544.1 |
POLR2B - RNA polymerase II subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000938.2 | Intron | NP_000929.1 | ||||
NM_001303268.1 | Intron | NP_001290197.1 | ||||
NM_001303269.1 | Intron | NP_001290198.1 |