Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCACCTTAGCCTCGAGGTCAGG[A/G]TCCTGAGGCCTGGTGTGTGTGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVA1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EVA1B - eva-1 homolog B (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304762.1 | Intron | NP_001291691.1 | ||||
NM_018166.2 | Intron | NP_060636.1 | ||||
XM_017001627.1 | Intron | XP_016857116.1 |
SH3D21 - SH3 domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162530.1 | Intron | NP_001156002.1 | ||||
NM_024676.4 | Intron | NP_078952.4 | ||||
XM_011542153.2 | Intron | XP_011540455.1 | ||||
XM_017002340.1 | Intron | XP_016857829.1 | ||||
XM_017002341.1 | Intron | XP_016857830.1 | ||||
XM_017002342.1 | Intron | XP_016857831.1 | ||||
XM_017002343.1 | Intron | XP_016857832.1 | ||||
XM_017002344.1 | Intron | XP_016857833.1 | ||||
XM_017002345.1 | Intron | XP_016857834.1 | ||||
XM_017002346.1 | Intron | XP_016857835.1 |