Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCTGAATACCTAGGAACTTTCA[A/G]GAAAACTAGCTTCTTAACAACAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601860 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD17B4 - hydroxysteroid 17-beta dehydrogenase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000414.3 | Intron | NP_000405.1 | ||||
NM_001199291.2 | Intron | NP_001186220.1 | ||||
NM_001199292.1 | Intron | NP_001186221.1 | ||||
NM_001292027.1 | Intron | NP_001278956.1 | ||||
NM_001292028.1 | Intron | NP_001278957.1 |