Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCAGTTTGCCTCCTCTAAAGAC[A/C]GACAAGATATTGGCTACCATGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118990 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSFA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SSFA2 - sperm specific antigen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130445.2 | 1478 | Intron | NP_001123917.1 | |||
NM_001287503.1 | 1478 | Intron | NP_001274432.1 | |||
NM_001287504.1 | 1478 | Intron | NP_001274433.1 | |||
NM_001287505.1 | 1478 | UTR 3 | NP_001274434.1 | |||
NM_006751.6 | 1478 | Intron | NP_006742.2 | |||
XM_005246812.1 | 1478 | Intron | XP_005246869.1 | |||
XM_005246813.1 | 1478 | Intron | XP_005246870.1 | |||
XM_011511701.2 | 1478 | Intron | XP_011510003.1 | |||
XM_011511702.2 | 1478 | Intron | XP_011510004.1 | |||
XM_011511703.2 | 1478 | Intron | XP_011510005.1 | |||
XM_017004782.1 | 1478 | Intron | XP_016860271.1 |