Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCAAGAATCTCTGCGGTTGCA[A/T]CTGAATTGCATCTAGTCACCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608998 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSSC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TSSC1 - tumor suppressing subtransferable candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006711893.2 | Intron | XP_006711956.1 | ||||
XM_011510383.2 | Intron | XP_011508685.1 | ||||
XM_011510384.2 | Intron | XP_011508686.1 | ||||
XM_011510385.2 | Intron | XP_011508687.1 | ||||
XM_011510387.2 | Intron | XP_011508689.1 | ||||
XM_017004818.1 | Intron | XP_016860307.1 |