Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146735 MIM: 601311 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGFBP6 - insulin like growth factor binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002178.2 | 299 | Intron | NP_002169.1 | |||
XM_017019264.1 | 299 | Intron | XP_016874753.1 |
SOAT2 - sterol O-acyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003578.3 | 299 | Intron | NP_003569.1 | |||
XM_011538849.2 | 299 | UTR 5 | XP_011537151.1 | |||
XM_011538850.2 | 299 | UTR 5 | XP_011537152.1 | |||
XM_011538851.2 | 299 | UTR 5 | XP_011537153.1 |
Set Membership: |
HapMap |