Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCTCCTGCAAACTTTGCAGGC[A/C]AGGTGGTAAGAATGAAGGCAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607568 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMAB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MMAB - methylmalonic aciduria (cobalamin deficiency) cblB type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052845.3 | 2056 | UTR 3 | NP_443077.1 | |||
XM_011538267.2 | 2056 | Intron | XP_011536569.1 | |||
XM_011538268.2 | 2056 | Intron | XP_011536570.1 | |||
XM_011538269.2 | 2056 | Intron | XP_011536571.1 |
Set Membership: |
HapMap |