Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTTTAGTGTAGGGTAGTAAAC[A/C]CAGCCAAAAGGGGTCGTTTATCTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603381 | |||||||||||||||||||||||
Literature Links: |
FLNB PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FLNB - filamin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164317.1 | Intron | NP_001157789.1 | ||||
NM_001164318.1 | Intron | NP_001157790.1 | ||||
NM_001164319.1 | Intron | NP_001157791.1 | ||||
NM_001457.3 | Intron | NP_001448.2 | ||||
XM_005264978.1 | Intron | XP_005265035.1 |