Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610533 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WWC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WWC1 - WW and C2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161661.1 | Intron | NP_001155133.1 | ||||
NM_001161662.1 | Intron | NP_001155134.1 | ||||
NM_015238.2 | Intron | NP_056053.1 | ||||
XM_005265850.1 | Intron | XP_005265907.1 | ||||
XM_005265853.2 | Intron | XP_005265910.1 | ||||
XM_011534485.1 | Intron | XP_011532787.1 | ||||
XM_011534486.1 | Intron | XP_011532788.1 | ||||
XM_011534487.1 | Intron | XP_011532789.1 | ||||
XM_011534488.1 | Intron | XP_011532790.1 | ||||
XM_011534489.1 | Intron | XP_011532791.1 | ||||
XM_011534490.1 | Intron | XP_011532792.1 | ||||
XM_011534491.1 | Intron | XP_011532793.1 | ||||
XM_017009276.1 | Intron | XP_016864765.1 | ||||
XM_017009277.1 | Intron | XP_016864766.1 | ||||
XM_017009278.1 | Intron | XP_016864767.1 |