Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAATCAGTCAAATGGAAAGGTT[C/T]TTAATCACTTAGGGGAATATCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609361 | ||||||||||||||||||||
Literature Links: |
HSPE1-MOB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPE1-MOB4 - HSPE1-MOB4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202485.1 | Intron | NP_001189414.1 |
MOB4 - MOB family member 4, phocein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100819.2 | Intron | NP_001094289.1 | ||||
NM_001204094.1 | Intron | NP_001191023.1 | ||||
NM_015387.4 | Intron | NP_056202.2 | ||||
NM_199482.3 | Intron | NP_955776.1 |