Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTATGGACTTTTCAGAAGGTG[C/T]CATATTTTAAATGTATCTGTTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HECW2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HECW2 - HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304840.1 | 8621 | Intron | NP_001291769.1 | |||
NM_020760.2 | 8621 | Intron | NP_065811.1 | |||
XM_006712646.3 | 8621 | UTR 3 | XP_006712709.1 | |||
XM_006712648.3 | 8621 | UTR 3 | XP_006712711.1 | |||
XM_017004563.1 | 8621 | UTR 3 | XP_016860052.1 |
Set Membership: |
HapMap |