Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGCATTAGGGAGGAGGGTCCTG[A/G]GGAAGTCAAAGCTTCCACACCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186921 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LMO1 - LIM domain only 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270428.1 | Intron | NP_001257357.1 | ||||
NM_002315.2 | Intron | NP_002306.1 | ||||
XM_006718228.3 | Intron | XP_006718291.1 | ||||
XM_011520098.2 | Intron | XP_011518400.1 | ||||
XM_011520099.2 | Intron | XP_011518401.1 | ||||
XM_011520100.1 | Intron | XP_011518402.1 |