Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATACAGTTTTTGTCACATTAGC[C/T]TCATGTAACTATTGCCACAATCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614657 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMOTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMOTL1 - angiomotin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301007.1 | Intron | NP_001287936.1 | ||||
NM_130847.2 | Intron | NP_570899.1 | ||||
XM_005273798.4 | Intron | XP_005273855.1 | ||||
XM_005273801.4 | Intron | XP_005273858.1 | ||||
XM_006718772.3 | Intron | XP_006718835.1 | ||||
XM_011542626.2 | Intron | XP_011540928.1 | ||||
XM_011542628.2 | Intron | XP_011540930.1 |
LOC105369438 - uncharacterized LOC105369438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |