Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATGGGTTCGTTCATCCAATTTC[C/T]ACCACTTTACAGATGAACTGGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01561 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC01561 - long intergenic non-protein coding RNA 1561 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLPP4 - phospholipid phosphatase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030059.2 | 9394 | Intron | NP_001025230.1 | |||
NM_001318166.1 | 9394 | Intron | NP_001305095.1 | |||
NM_001318167.1 | 9394 | Intron | NP_001305096.1 | |||
NM_001318168.1 | 9394 | Intron | NP_001305097.1 | |||
NM_001318169.1 | 9394 | Intron | NP_001305098.1 | |||
XM_005269592.2 | 9394 | UTR 3 | XP_005269649.1 | |||
XM_006717685.3 | 9394 | UTR 3 | XP_006717748.1 | |||
XM_006717686.3 | 9394 | Intron | XP_006717749.1 | |||
XM_011539444.2 | 9394 | UTR 3 | XP_011537746.1 | |||
XM_011539445.2 | 9394 | Intron | XP_011537747.1 | |||
XM_017015820.1 | 9394 | Intron | XP_016871309.1 | |||
XM_017015821.1 | 9394 | UTR 3 | XP_016871310.1 | |||
XM_017015822.1 | 9394 | UTR 3 | XP_016871311.1 | |||
XM_017015823.1 | 9394 | Intron | XP_016871312.1 | |||
XM_017015824.1 | 9394 | Intron | XP_016871313.1 |
Set Membership: |
HapMap |