Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGCCATGAGGTTGTGCAGCTG[A/C]GTTATGATGCTCTAGGGCTGAGGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RRP12 - ribosomal RNA processing 12 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145114.1 | Intron | NP_001138586.1 | ||||
NM_001284337.1 | Intron | NP_001271266.1 | ||||
NM_015179.3 | Intron | NP_055994.2 | ||||
XM_011539555.1 | Intron | XP_011537857.1 | ||||
XM_011539556.2 | Intron | XP_011537858.1 |