Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGGTCAAATACTCTGATGTCA[C/G]ACCCAAGAAATAATAAAAAATGTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611587 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP19 - Rho GTPase activating protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204300.1 | 2084 | UTR 3 | NP_001191229.1 | |||
NM_001256423.1 | 2084 | UTR 3 | NP_001243352.1 | |||
NM_032900.5 | 2084 | UTR 3 | NP_116289.4 |
ARHGAP19-SLIT1 - ARHGAP19-SLIT1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |