Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCAGCACTGGCTCATACTGATT[A/C]ATTTTGATCTCTGCTAATACCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136130 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FMO1 - flavin containing monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282692.1 | 82 | Intron | NP_001269621.1 | |||
NM_001282693.1 | 82 | UTR 5 | NP_001269622.1 | |||
NM_001282694.1 | 82 | UTR 5 | NP_001269623.1 | |||
NM_002021.2 | 82 | UTR 5 | NP_002012.1 | |||
XM_005245037.3 | 82 | Intron | XP_005245094.1 | |||
XM_005245038.3 | 82 | UTR 5 | XP_005245095.1 | |||
XM_006711241.3 | 82 | UTR 5 | XP_006711304.1 | |||
XM_006711242.3 | 82 | UTR 5 | XP_006711305.1 |
Set Membership: |
HapMap DME Validated Inventoried |