Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTGGAAGCCTCTGTATTATCTG[G/T]CAAGATGGTAACCAAACTTAGAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605667 | ||||||||||||||||||||
Literature Links: |
COLGALT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COLGALT2 - collagen beta(1-O)galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303420.1 | Intron | NP_001290349.1 | ||||
NM_001303421.1 | Intron | NP_001290350.1 | ||||
NM_015101.3 | Intron | NP_055916.1 | ||||
XM_011509332.2 | Intron | XP_011507634.1 | ||||
XM_011509334.2 | Intron | XP_011507636.1 | ||||
XM_011509335.2 | Intron | XP_011507637.1 | ||||
XM_011509336.2 | Intron | XP_011507638.1 | ||||
XM_011509337.2 | Intron | XP_011507639.1 | ||||
XM_011509338.2 | Intron | XP_011507640.1 | ||||
XM_017000750.1 | Intron | XP_016856239.1 | ||||
XM_017000751.1 | Intron | XP_016856240.1 |
RGL1 - ral guanine nucleotide dissociation stimulator like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |