Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACTGATTTCTTATGAGTGAAG[A/G]AAGAACGTGTTAATCGGCAGGAGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609335 MIM: 189940 | |||||||||||||||||||||||
Literature Links: |
C1orf27 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
C1orf27 - chromosome 1 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164245.1 | Intron | NP_001157717.1 | ||||
NM_001164246.1 | Intron | NP_001157718.1 | ||||
NM_017847.5 | Intron | NP_060317.3 | ||||
XM_011509660.1 | Intron | XP_011507962.1 | ||||
XM_011509661.1 | Intron | XP_011507963.1 | ||||
XM_017001543.1 | Intron | XP_016857032.1 |
TPR - translocated promoter region, nuclear basket protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |