Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTACTGAATATCCCTTGTCTTGT[C/T]CTCCTCCTCCCCAAACTTAATGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600160 MIM: 600431 MIM: 613149 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDKN2A - cyclin dependent kinase inhibitor 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKN2B - cyclin dependent kinase inhibitor 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKN2B-AS1 - CDKN2B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |