Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAGTTTTGTTTCTTTTGACACT[A/G]TAAAATTTTGAGATTCTAAATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603785 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPDZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPDZ - multiple PDZ domain crumbs cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261406.1 | Intron | NP_001248335.1 | ||||
NM_001261407.1 | Intron | NP_001248336.1 | ||||
NM_003829.4 | Intron | NP_003820.2 | ||||
XM_005251622.4 | Intron | XP_005251679.1 | ||||
XM_005251623.4 | Intron | XP_005251680.1 | ||||
XM_006716885.3 | Intron | XP_006716948.1 | ||||
XM_006716886.3 | Intron | XP_006716949.1 | ||||
XM_006716887.3 | Intron | XP_006716950.1 | ||||
XM_006716888.3 | Intron | XP_006716951.1 | ||||
XM_006716889.3 | Intron | XP_006716952.1 | ||||
XM_006716891.3 | Intron | XP_006716954.1 | ||||
XM_017015252.1 | Intron | XP_016870741.1 | ||||
XM_017015253.1 | Intron | XP_016870742.1 | ||||
XM_017015254.1 | Intron | XP_016870743.1 | ||||
XM_017015255.1 | Intron | XP_016870744.1 | ||||
XM_017015256.1 | Intron | XP_016870745.1 | ||||
XM_017015257.1 | Intron | XP_016870746.1 | ||||
XM_017015258.1 | Intron | XP_016870747.1 | ||||
XM_017015259.1 | Intron | XP_016870748.1 |