Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGATGTATATGTGTGCATGCAC[A/G]TCTTTAAAAAACACCAACGGCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616618 MIM: 608221 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACBD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MASTL - microtubule associated serine/threonine kinase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172303.2 | Intron | NP_001165774.1 | ||||
NM_001172304.2 | Intron | NP_001165775.1 | ||||
NM_001320756.1 | Intron | NP_001307685.1 | ||||
NM_001320757.1 | Intron | NP_001307686.1 | ||||
NM_032844.4 | Intron | NP_116233.2 | ||||
XM_005252631.3 | Intron | XP_005252688.1 | ||||
XM_005252632.3 | Intron | XP_005252689.1 | ||||
XM_006717519.3 | Intron | XP_006717582.1 | ||||
XM_006717520.3 | Intron | XP_006717583.1 | ||||
XM_011519751.2 | Intron | XP_011518053.1 | ||||
XM_017016852.1 | Intron | XP_016872341.1 | ||||
XM_017016853.1 | Intron | XP_016872342.1 | ||||
XM_017016854.1 | Intron | XP_016872343.1 | ||||
XM_017016855.1 | Intron | XP_016872344.1 | ||||
XM_017016856.1 | Intron | XP_016872345.1 |