Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179020 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDXK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDXK - pyridoxal (pyridoxine, vitamin B6) kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003681.4 | Intron | NP_003672.1 | ||||
XM_005261195.2 | Intron | XP_005261252.2 | ||||
XM_005261196.3 | Intron | XP_005261253.1 | ||||
XM_005261198.2 | Intron | XP_005261255.1 | ||||
XM_005261199.2 | Intron | XP_005261256.1 | ||||
XM_011529758.2 | Intron | XP_011528060.1 | ||||
XM_011529760.2 | Intron | XP_011528062.1 | ||||
XM_011529761.1 | Intron | XP_011528063.1 | ||||
XM_011529762.2 | Intron | XP_011528064.1 | ||||
XM_017028483.1 | Intron | XP_016883972.1 | ||||
XM_017028484.1 | Intron | XP_016883973.1 |
Set Membership: |
HapMap |