Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAGGCAGGGCTGTCTGCACCCA[C/T]TGAGACCTCCAGCAGCATCCCCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609626 | ||||||||||||||||||||
Literature Links: |
MDGA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MDGA1 - MAM domain containing glycosylphosphatidylinositol anchor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153487.3 | 5642 | UTR 3 | NP_705691.1 | |||
XM_006715056.3 | 5642 | Intron | XP_006715119.1 | |||
XM_017010734.1 | 5642 | UTR 3 | XP_016866223.1 |