Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAGCCCGTGTGTAAAACGCAGA[C/T]GGTCACATCCGCCTTCCAGAACAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606378 MIM: 610135 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ST6GALNAC4 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175039.3 | Intron | NP_778204.1 | ||||
NM_175040.3 | Intron | NP_778205.1 | ||||
XM_011518527.2 | Intron | XP_011516829.1 | ||||
XM_011518529.2 | Intron | XP_011516831.1 |
ST6GALNAC6 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |