Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCGGCGCGGGCATGGGCGCGTG[C/T]TATGACGGCGCAGGGCGCCCGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604349 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAMC3 - laminin subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006059.3 | 173 | Silent Mutation | TGC,TGT | C,C 25 | NP_006050.3 | |
XM_006716921.2 | 173 | Silent Mutation | TGC,TGT | C,C 25 | XP_006716984.1 | |
XM_011518121.1 | 173 | Silent Mutation | TGC,TGT | C,C 25 | XP_011516423.1 |
Set Membership: |
HapMap |