Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGGCTATTTTAATGAAAGAACA[C/T]TTAAAAATAACATCAGAGAGATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601330 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf60 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf60 - chromosome 12 open reading frame 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175874.3 | Intron | NP_787070.2 | ||||
XM_005253322.4 | Intron | XP_005253379.1 | ||||
XM_011520568.2 | Intron | XP_011518870.1 | ||||
XM_011520569.2 | Intron | XP_011518871.1 | ||||
XM_017018871.1 | Intron | XP_016874360.1 | ||||
XM_017018872.1 | Intron | XP_016874361.1 | ||||
XM_017018873.1 | Intron | XP_016874362.1 | ||||
XM_017018874.1 | Intron | XP_016874363.1 |
GUCY2C - guanylate cyclase 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004963.3 | Intron | NP_004954.2 | ||||
XM_011520631.2 | Intron | XP_011518933.1 |
PLBD1-AS1 - PLBD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |