Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAATCCAGTGTGGCTTAGGAA[C/T]AGGACACTAGACTGGGGATCAGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615079 MIM: 608858 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FGFR1OP2 - FGFR1 oncogene partner 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171887.1 | Intron | NP_001165358.1 | ||||
NM_001171888.1 | Intron | NP_001165359.1 | ||||
NM_015633.2 | Intron | NP_056448.1 |
Set Membership: |
HapMap |