Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGTTATTTTTCTTATTCCTGGG[G/T]TACAACGAAGGAATGTGAGTTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616836 MIM: 611032 | ||||||||||||||||||||
Literature Links: |
GPATCH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPATCH2 - G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA17 - spermatogenesis associated 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138796.3 | Intron | NP_620151.1 | ||||
XM_005273052.1 | Intron | XP_005273109.1 | ||||
XM_006711165.3 | Intron | XP_006711228.1 | ||||
XM_011509194.2 | Intron | XP_011507496.1 |