Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTATTCTTTGATTCTTTGTTGT[A/G]CTATTAACATGACTTTCTACCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPATCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPATCH2 - G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297754.1 | 5292 | Intron | NP_001284683.1 | |||
NM_018040.3 | 5292 | UTR 3 | NP_060510.1 | |||
XM_011509689.2 | 5292 | Intron | XP_011507991.1 | |||
XM_011509690.2 | 5292 | Intron | XP_011507992.1 | |||
XM_011509691.2 | 5292 | Intron | XP_011507993.1 | |||
XM_011509693.2 | 5292 | Intron | XP_011507995.1 | |||
XM_011509694.2 | 5292 | Intron | XP_011507996.1 | |||
XM_017001592.1 | 5292 | Intron | XP_016857081.1 | |||
XM_017001593.1 | 5292 | Intron | XP_016857082.1 |