Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTTTCTTTATATATGTGGACG[C/T]CTATCCTGGTTTGTTTTATCAGGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603497 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYRK3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DYRK3 - dual specificity tyrosine phosphorylation regulated kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004023.1 | Intron | NP_001004023.1 | ||||
NM_003582.2 | Intron | NP_003573.2 | ||||
XM_005273315.4 | Intron | XP_005273372.1 | ||||
XM_011510061.2 | Intron | XP_011508363.1 |