Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCAAGTTCAGCCACCAGGCCC[C/G]TTAAGCACAGTGCCAAGAGCCTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM186B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM186B - family with sequence similarity 186 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032130.2 | 3159 | Intron | NP_115506.1 | |||
XM_006719625.2 | 3159 | UTR 3 | XP_006719688.1 | |||
XM_006719626.2 | 3159 | Missense Mutation | ACG,AGG | T,R 852 | XP_006719689.1 | |
XM_006719627.3 | 3159 | Intron | XP_006719690.1 | |||
XM_011538796.2 | 3159 | Intron | XP_011537098.1 | |||
XM_011538797.2 | 3159 | Missense Mutation | AAC,AAG | N,K 795 | XP_011537099.1 | |
XM_017020008.1 | 3159 | UTR 3 | XP_016875497.1 |