Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGCTTAAGGTCATAGAGCAAGAT[A/G]TAGATGAGGTGGGATTCAAACCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615089 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG101 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATG101 - autophagy related 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098673.1 | Intron | NP_001092143.1 | ||||
NM_021934.4 | Intron | NP_068753.2 |
OR7E47P - olfactory receptor family 7 subfamily E member 47 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |