Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCTACCTTCTGCTCTCATCCT[A/G]CCTAAAAGTCATAGAAAAACGTTGA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 605257 | |||||||||||||||||||||||||||||
Literature Links: |
AICDA PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
AICDA - activation-induced cytidine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011520772.2 | 1074 | UTR 3 | XP_011519074.1 | |||
XM_011520773.2 | 1074 | Intron | XP_011519075.1 |
Set Membership: |
HapMap |