Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTTGGAGGTAAAAGAGACAATA[A/C]CAGGAGAAGAAAACTACATAAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300157 | ||||||||||||||||||||
Literature Links: |
ACSL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSL4 - acyl-CoA synthetase long-chain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318509.1 | Intron | NP_001305438.1 | ||||
NM_001318510.1 | Intron | NP_001305439.1 | ||||
NM_004458.2 | Intron | NP_004449.1 | ||||
NM_022977.2 | Intron | NP_075266.1 | ||||
XM_005262109.2 | Intron | XP_005262166.1 | ||||
XM_006724635.2 | Intron | XP_006724698.1 | ||||
XM_011530888.2 | Intron | XP_011529190.1 | ||||
XM_011530889.2 | Intron | XP_011529191.1 |